Proteintech
Manufacturer Code:207411AP150UL
Catalog # 50-173-3058
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Brachyury protein; Brachyury protein Brachyury T Protein T T T brachyury homolog (mouse) TFT; brachyury-like 2; brachyury-like 3; low ratio; Protein T; T brachyury homolog; T, brachyury homolog; T-box transcription factor T
Gene Aliases: Bra; cou; D17Mit170; Low; Lr; me75; SAVA; T; T1; TBXT; TFT; Tl2; Tl3
UniProt ID: (Human) O15178, (Mouse) P20293
Entrez Gene ID: (Human) 6862, (Mouse) 20997
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.