Novus Biologicals
Manufacturer Code:NB10093545SS
Catalog # NB001051
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to an internal region of human Brachyury. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
Protein Aliases: Brachyury protein; brachyury protein MGC104817 Protein T SAVA T brachyury (mouse) homolog T brachyury homolog T Brachyury Transcription Factor T brachyury homolog (mouse) T-Box Transcription Factor T TBXT TF TFT; Protein T; T brachyury homolog; T, brachyury homolog; T-box transcription factor T
Gene Aliases: SAVA; T; TBXT; TFT
UniProt ID: (Human) O15178
Entrez Gene ID: (Human) 6862
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.