R&D Systems
Manufacturer Code:AF2085
Catalog # AF2085
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Brachyury Ser2-Glu202 Accession # O15178 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Brachyury protein; brachyury protein MGC104817 Protein T SAVA T brachyury (mouse) homolog T brachyury homolog T Brachyury Transcription Factor T brachyury homolog (mouse) T-Box Transcription Factor T TBXT TF TFT; Protein T; T brachyury homolog; T, brachyury homolog; T-box transcription factor T
Gene Aliases: SAVA; T; TBXT; TFT
UniProt ID: (Human) O15178
Entrez Gene ID: (Human) 6862
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.