Novus Biologicals
Manufacturer Code:NB100214H
Catalog # NB100214H
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 1425 and the C-terminus (residue 1477) of human Bloom Syndrome using the numbering given in entry NP_000048.1 (GeneID 641). |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bloom syndrome Bloom syndrome RecQ helicase-like BSBloom syndrome protein DNA helicase RecQ-like type 2 EC 3.6.1 EC 3.6.4.12 MGC126616 MGC131618 RecQ protein-like 3 RecQ2 RECQ2MGC131620 RECQL3RECQL2; Bloom syndrome protein; Bloom syndrome, RecQ helicase-like; DNA helicase, RecQ-like type 2; RecQ protein-like 3; RecQ2
Gene Aliases: BLM; BS; RECQ2; RECQL2; RECQL3
UniProt ID: (Human) P54132
Entrez Gene ID: (Human) 641
Molecular Function:
DNA binding protein
DNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.