Invitrogen
Manufacturer Code:PA585140
Catalog # PIPA585140
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the N-terminus region of human BLM. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bloom syndrome protein; Bloom syndrome, RecQ helicase-like; BS antibody MGC126616 antibody MGC131618 antibody MGC131620 antibody RECQ2 antibody RECQL2 antibody RECQL3 antibody BLM antibody DNA helicase RecQ-like type 2 antibody Bloom syndrome protein antibody recQ protein-like 3 antibody Bloom syndrome RecQ helicase-like antibody; DNA helicase, RecQ-like type 2; RecQ protein-like 3; RecQ2
Gene Aliases: BLM; BS; RECQ2; RECQL2; RECQL3
UniProt ID: (Human) P54132
Entrez Gene ID: (Human) 641
Molecular Function: DNA binding protein DNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.