Invitrogen
Manufacturer Code:12985080
Catalog # 50-112-2467
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 5E7 |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: B lymphocyte-induced maturation protein 1; Beta-interferon gene positive regulatory domain I-binding factor; BLIMP 1 B Lymphocyte Induced Maturation Protein 1 BLIMP1 Positive Regulatory Domain I Binding Factor 1 PR Domain Containing 1 PRDI BF1 PRDM1; Blimp-1; PR domain zinc finger protein 1; PR domain-containing protein 1
Gene Aliases: b2b1765Clo; Blimp-1; Blimp1; PRDI-BF1; Prdm1; ZNFPR1A1
UniProt ID: (Mouse) Q60636
Entrez Gene ID: (Mouse) 12142
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.