Test
Invitrogen
Manufacturer Code:17985080
Catalog # 17-985-080
Quantity:
Shared lists are a new way to save products. Create a list for each of your projects to share with collaborators like your lab manager. Then select and buy products as a team.
![]()
Creating a shared list
Click "Save to list" to create your first list, give it a name, and add the product. Access your shared lists in the Account link on Thermofisher.com. |
![]()
Collaborate
You can invite collaborators like your lab manager or procurement staff to any shared list. They can add more items, leave comments and complete the final purchase. |
![]()
Export as a spreadsheet
Export your list as a spreadsheet for easy documentation or hand off to your lab’s designated buyer. |
Shared lists are a new way to save products. Create a list for each of your projects to share with collaborators like your lab manager. Then select and buy products as a team.
Click "Save to list" to create your first list, give it a name, and add the product. Access your shared lists in the Account link on Thermofisher.com.
You can invite collaborators like your lab manager or procurement staff to any shared list. They can add more items, leave comments and complete the final purchase.
Export your list as a spreadsheet for easy documentation or hand off to your lab’s designated buyer.
Shared lists are a new way to save products. Create a list for each of your projects to share with collaborators like your lab manager. Then select and buy products as a team.
![]()
Creating a shared list
Click "Save to list" to create your first list, give it a name, and add the product. Access your shared lists in the Account link on Thermofisher.com. |
![]()
Collaborate
You can invite collaborators like your lab manager or procurement staff to any shared list. They can add more items, leave comments and complete the final purchase. |
Export your list as a spreadsheet for easy documentation or hand off to your lab’s designated buyer.
Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 5E7 |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4°C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: B lymphocyte-induced maturation protein 1; B lymphocyte-induced maturation protein 1 b2b1765Clo Beta-interferon gene positive regulatory domain I-binding factor beta-interferon gene positive-regulatory domain I binding factor BLIMP 1 Blimp1 BLIMP-1 B-lymphocyte-induced maturation protein 1 MGC118922 MGC118923 MGC118924 MGC118925 OTTHUMP00000016918 OTTHUMP00000202173 Positive regulatory domain I-binding factor 1 PR domain 1 PR domain containing 1 with ZNF domain PR domain zinc finger protein 1 PR domain-containing protein 1 PRDI BF1 PRDI-BF1 PRDI-binding factor-1 Prdm1 RP1-134E15.1 ZNFPR1A1; Beta-interferon gene positive regulatory domain I-binding factor; Blimp-1; PR domain zinc finger protein 1; PR domain-containing protein 1
Gene Aliases: b2b1765Clo; Blimp-1; Blimp1; PRDI-BF1; Prdm1; ZNFPR1A1
UniProt ID: (Mouse) Q60636
Entrez Gene ID: (Mouse) 12142
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.