Invitrogen
Manufacturer Code:MA516116
Catalog # PIMA516116
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3H2-E8 |
Immunogen | A fragment of the mouse Blimp-1 (residues 199-409). |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: B lymphocyte-induced maturation protein 1; B-lymphocyte-induced maturation protein 1; Beta-interferon gene positive regulatory domain I-binding factor; beta-interferon gene positive-regulatory domain I binding factor; BLIMP-1; Positive regulatory domain I-binding factor 1; PR domain containing 1, with ZNF domain; PR domain zinc finger protein 1; PR domain-containing protein 1; PRDI-BF1; PRDI-binding factor-1; RP1-134E15.1 BLIMP1 PRDI-BF1 B-lymphocyte-induced maturation protein 1 BLIMP-1 PR domain zinc finger protein 1 PRDI-binding factor-1 beta-interferon gene positive-regulatory domain I binding factor
Gene Aliases: b2b1765Clo; Blimp-1; BLIMP1; PRDI-BF1; PRDM1; ZNFPR1A1
UniProt ID: (Human) O75626, (Mouse) Q60636
Entrez Gene ID: (Pig) 100154284, (Human) 639, (Mouse) 12142
Molecular Function:
DNA binding protein
centromere DNA-binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.