Invitrogen
Manufacturer Code:PA518110
Catalog # PIPA518110
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (KVKQETVEPMDP) corresponding to the C-terminus amino acids of PRDM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: B-lymphocyte-induced maturation protein 1; Beta-interferon gene positive regulatory domain I-binding factor; beta-interferon gene positive-regulatory domain I binding factor; BLIMP-1; Positive regulatory domain I-binding factor 1; PR domain containing 1, with ZNF domain; PR domain zinc finger protein 1; PR domain-containing protein 1; PRDI-BF1; PRDI-binding factor-1; PRDM1 PR domain containing 1 with ZNF domain RP1-134E15.1 BLIMP1 MGC118922 MGC118923 MGC118924 MGC118925 PRDI-BF1 B-lymphocyte-induced maturation protein 1 PR domain containing 1 with ZNF domain isoform 2 PR-domain zinc finger protein 1 PRDI-binding factor-1 beta-interferon gene positive-regulatory domain I binding factor positive regulatory domain I-binding factor 1
Gene Aliases: BLIMP1; PRDI-BF1; PRDM1
UniProt ID: (Human) O75626
Entrez Gene ID: (Human) 639
Molecular Function:
DNA binding protein
centromere DNA-binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.