Invitrogen
Manufacturer Code:PA520168
Catalog # PIPA520168
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 16 amino acid peptide from near the center of human Bit1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bcl-2 inhibitor of transcription 1; CGI-147 BIT1 PTH2 PTH 2 bcl-2 inhibitor of transcription 1 peptidyl-tRNA hydrolase 2 mitochondrial; cilia and flagella associated protein 37; Peptidyl-tRNA hydrolase 2, mitochondrial; PTH 2
Gene Aliases: A230072I16Rik; BIT1; CFAP37; CGI-147; IMNEPD; PTH; PTH 2; PTH2; PTRH2
UniProt ID: (Human) Q9Y3E5, (Mouse) Q8R2Y8
Entrez Gene ID: (Human) 51651, (Mouse) 217057
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.