Invitrogen
Manufacturer Code:PA517327
Catalog # PIPA517327
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues within a central region of Bit1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bcl-2 inhibitor of transcription 1; cilia and flagella associated protein 37; OTTHUMP00000205936 PTH 2 bcl-2 inhibitor of transcription 1 peptidyl-tRNA hydrolase 2 mitochondrial; Peptidyl-tRNA hydrolase 2, mitochondrial; PTH 2
Gene Aliases: BIT1; CFAP37; CGI-147; IMNEPD; PTH; PTH 2; PTH2; PTRH2
UniProt ID: (Human) Q9Y3E5
Entrez Gene ID: (Human) 51651
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.