Invitrogen
Manufacturer Code:PA553434
Catalog # PIPA553434
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human Bisphosphoglycerate mutase |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2,3-bisphosphoglycerate mutase; 2,3-bisphosphoglycerate mutase, erythrocyte; 2,3-bisphosphoglycerate synthase; 2,3-diphosphoglycerate mutase; Bisphosphoglycerate mutase; BPG-dependent PGAM; BPGM; DPGM; DPGM 23-bisphosphoglycerate mutase erythrocyte 23-bisphosphoglycerate synthase 23-diphosphoglycerate mutase BPG-dependent PGAM bisphosphoglycerate mutase erythrocyte 23-bisphosphoglycerate mutase; erythrocyte 2,3-bisphosphoglycerate mutase; testis secretory sperm-binding protein Li 202a
Gene Aliases: Ab2-098; AI323730; AL022789; BPGM; C86192; DPGM
UniProt ID: (Human) P07738, (Mouse) P15327
Entrez Gene ID: (Human) 669, (Rat) 296973, (Mouse) 12183
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.