Invitrogen
Manufacturer Code:MA514848
Catalog # PIMA514848
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | K.912.7 |
Immunogen | Synthetic peptide corresponding to residues surrounding Pro25 of Bim |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: bcl-2 interacting mediator of cell death; bcl-2 interacting protein Bim; Bcl-2 related apoptotic gene product BimL; Bcl-2-like protein 11; bcl-2-like protein 11-like; bcl-2-related ovarian death agonist; Bcl-2-related ovarian death protein; Bcl2 interacting mediator of cell death; Bcl2-interacting mediator of cell death; Bcl2-L-11; BCL2-like 11 (apoptosis facilitator); OTTHUMP00000197013 OTTHUMP00000204247 bcl-2 interacting mediator of cell death bcl-2 interacting protein Bim bcl-2-like protein 11 bcl-2-related ovarian death agonist bcl2-L-11 bcl2-interacting mediator of cell death
Gene Aliases: 1500006F24Rik; BAM; bcl2-L-11; BCL2L11; BIM; BIMEL; BimL; BOD
UniProt ID: (Human) O43521, (Mouse) O54918, (Rat) O88498
Entrez Gene ID: (Dog) 612867, (Human) 10018, (Bovine) 507390, (Pig) 396632, (Mouse) 12125, (Rat) 64547
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.