Novus Biologicals
Manufacturer Code:NBP175511
Catalog # NBP175511
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1C2H4 |
Immunogen | Antibody was raised against a recombinant protein corresponding to amino acids 2 to 198 of human Bim. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20 degrees C. Avoid freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BAM bcl-2 interacting mediator of cell death bcl-2 interacting protein Bim Bcl2-interacting mediator of cell death bcl2-L-11 BCL2-like 11 (apoptosis facilitator) bcl-2-like protein 11 bcl-2-related ovarian death agonist BIM-alpha6 BIM-beta6 BIM-beta7 BIMBOD BimEL BimL; bcl-2 interacting mediator of cell death; bcl-2 interacting protein Bim; Bcl-2-like protein 11; bcl-2-related ovarian death agonist; Bcl2-interacting mediator of cell death; Bcl2-L-11; BCL2-like 11 (apoptosis facilitator)
Gene Aliases: BAM; BCL2L11; BIM; BOD
UniProt ID: (Human) O43521
Entrez Gene ID: (Human) 10018
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.