Novus Biologicals
Manufacturer Code:NB10062870
Catalog # NB10062870
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 4 -17 (A-S-G-A-D-T-S-G-V-L-D-P-D-S) of the native molecule conjugated to Keyhole limpet Haemocyanin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: Biglycan; biglycan Bone/cartilage proteoglycan I bone/cartilage proteoglycan-I dermatan sulphate proteoglycan I DSPG1 PGI PG-S1 SLRR1Abiglycan proteoglycan small leucine-rich protein 1A; Bone/cartilage proteoglycan I; bone/cartilage proteoglycan-I; dermatan sulphate proteoglycan I; PG-S1; small leucine-rich protein 1A
Gene Aliases: BGN; DSPG1; PG-S1; PGI; SLRR1A
UniProt ID: (Human) P21810
Entrez Gene ID: (Human) 633
Molecular Function: cytokine extracellular matrix protein receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.