Invitrogen
Manufacturer Code:PA528068
Catalog # PIPA528068
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 302 and 365 of Human Bif1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bax-interacting factor 1; Bif-1; Bif-1 PPP1R70 dJ612B15.2 Bax-interacting factor 1 SH3 domain-containing GRB2-like protein B1 SH3-containing protein SH3GLB1 endophilin-B1 protein phosphatase 1 regulatory subunit 70; Endophilin-B1; protein phosphatase 1, regulatory subunit 70; SH3 domain-containing GRB2-like protein B1; SH3-containing protein SH3GLB1; SH3-domain GRB2 like endophilin B1; SH3-domain GRB2-like endophilin B1; testicular tissue protein Li 172
Gene Aliases: AA409932; AI314629; AU015566; Bif-1; CGI-61; dJ612B15.2; KIAA0491; mKIAA0491; PPP1R70; SH3GLB1
UniProt ID: (Human) Q9Y371, (Mouse) Q9JK48
Entrez Gene ID: (Human) 51100, (Mouse) 54673
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.