Invitrogen
Manufacturer Code:MA516225
Catalog # PIMA516225
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 20F565 |
Immunogen | A mixture of three synthetic peptides corresponding to the N-terminal internal and C-terminal regions of human B1. Epitope mapping indicates that the antibody recognizes an epitope in the N-terminal region of Bi-1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Bax inhibitor 1; Bax inhibitor-1; BAXI1 BI-1 TEGT bax inhibitor 1 testis enhanced gene transcript testis-enhanced gene transcript protein transmembrane BAX inhibitor motif-containing protein 6; BI-1; testis enhanced gene transcript; Testis-enhanced gene transcript protein; Transmembrane BAX inhibitor motif-containing protein 6
Gene Aliases: 5031406P05Rik; Ab1-011; Ac1-149; BAXI1; BI-1; BI1; Cc1-27; TEGT; TMBIM6
UniProt ID: (Human) P55061, (Rat) P55062, (Mouse) Q9D2C7
Entrez Gene ID: (Human) 7009, (Rat) 24822, (Mouse) 110213
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.