Proteintech
Manufacturer Code:155181AP150UL
Catalog # 50-173-0282
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Beta galactosidase Fusion Protein Ag7792 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acid beta galactosidase Beta galactosidase EBP Elastin receptor 1 ELNR1 galactosidase beta 1 GLB1 Lactase; Acid beta-galactosidase; Beta-galactosidase; Elastin receptor 1; elastin receptor 1, 67kDa; Lactase
Gene Aliases: AW125515; Bge; Bgl; Bgl-e; Bgl-s; Bgl-t; Bgs; Bgt; C130097A14Rik; EBP; ELNR1; Glb-1; GLB1; MPS4B
UniProt ID: (Human) P16278, (Mouse) P23780
Entrez Gene ID: (Human) 2720, (Mouse) 12091
Molecular Function:
galactosidase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.