OriGene
Manufacturer Code:TA502304
Catalog # 50-167-3369
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1E10 |
Immunogen | Full length human recombit protein of human CTNNB1 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Beta-catenin; catenin (cadherin-associated protein), beta 1; catenin (cadherin-associated protein), beta 1, 88kDa; Catenin beta-1; CTNNB EVR7 MRD19 armadillo
Gene Aliases: armadillo; CTNNB; CTNNB1; MRD19; OK/SW-cl.35; PRO2286
UniProt ID: (Dog) B6V8E6, (Human) P35222
Entrez Gene ID: (Dog) 477032, (Human) 1499
Molecular Function:
cell adhesion molecule
cytoskeletal protein
signaling molecule
storage protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.