Novus Biologicals
Manufacturer Code:NBP132353
Catalog # NBP132353
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human HSD3B2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3 beta-HSD type II; 3 beta-HSD type II 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 2 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type II 3-beta-HSD II delta 5-delta 4-isomerase type II HSD3B HSDB HSDB3B hydroxy-delta-5-steroid dehydrogenase 3 beta- and steroid delta-isomerase 23-beta-hydroxy-Delta(5)-steroid dehydrogenase progesterone reductase SDR11E2 short chain dehydrogenase/reductase family 11E member 23-beta-hydroxy-5-ene steroid dehydrogenase; 3 beta-hydroxysteroid dehydrogenase type II, delta 5-delta 4-isomerase type II, 3 beta-HSD type II; 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 2; 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type II; 3-beta-HSD adrenal and gonadal type; 3-beta-HSD II; 3-beta-hydroxy-5-ene steroid dehydrogenase; 3-beta-hydroxy-Delta(5)-steroid dehydrogenase; delta 5-delta 4-isomerase type II; Delta-5-3-ketosteroid isomerase; Progesterone reductase; short chain dehydrogenase/reductase family 11E, member 2; Steroid Delta-isomerase
Gene Aliases: HSD3B; HSD3B2; HSDB; HSDB3B; SDR11E2
UniProt ID: (Human) P26439
Entrez Gene ID: (Human) 3284
Molecular Function: dehydratase epimerase/racemase isomerase lyase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.