Novus Biologicals
Manufacturer Code:NBP29219920UL
Catalog # NB026290
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 30-200 of human B3GNT6 (NP_619651.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetylgalactosaminyl-O-glycosyl-glycoprotein beta-1,3-N-acetylglucosaminyltransferase; B3Gn-T6 beta-13-Gn-T6 Beta-13-N-acetylglucosaminyltransferase 6 beta-13-N-acetylglucosaminyltransferase protein beta3Gn-T6 BGnT-6 Core 3 synthase EC 2.4.1.- IMAGE:4907098 MGC119334 MGC119336 MGC119337 UDP-GlcNAc:betaGal beta-13-N-acetylglucosaminyltransferase 6 UDP-GlcNAc:betaGal beta-13-N-acetylglucosaminyltransferase 6 (core 3 synthase); beta-1,3-N-acetylglucosaminyltransferase 6; beta-1,3-N-acetylglucosaminyltransferase protein; BGnT-6; Core 3 synthase; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6
Gene Aliases: B3Gn-T6; B3GNT6; beta-1,3-Gn-T6; beta3Gn-T6; BGnT-6
UniProt ID: (Human) Q6ZMB0
Entrez Gene ID: (Human) 192134
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.