R&D Systems
Manufacturer Code:AF6664
Catalog # AF6664
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human beta-14-Glucuronyltransferase 1/B4GAT1 Asp43-Cys415 Accession # O43505 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: B3GN-T1 beta-13-N-acetylglucosaminyltransferase bGnT-6 BETA3GNTI EC 2.4.1.149 i-beta-13-N-acetylglucosaminyltransferase iGAT IGNT N-acetyllactosaminide beta-13-N-acetylglucosaminyltransferase Poly-N-acetyllactosamine extension enzyme UDP-GlcNAc:betaGal beta-13-N-acetylglucosaminyltransferase 1B3GNT6 UDP-GlcNAc:betaGal beta-13-N-acetylglucosaminyltransferase 6; beta-1,3-N-acetylglucosaminyltransferase bGnT-6; Beta-1,4-glucuronyltransferase 1; I-beta-1,3-N-acetylglucosaminyltransferase; iGnT; N-acetyllactosaminide beta-1,3-N-acetylglucosaminyltransferase; Poly-N-acetyllactosamine extension enzyme; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 1; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6
Gene Aliases: B3GN-T1; B3GNT1; B3GNT6; B4GAT1; BETA3GNTI; iGAT; iGNT; MDDGA13
UniProt ID: (Human) O43505
Entrez Gene ID: (Human) 11041
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.