Invitrogen
Manufacturer Code:PA569670
Catalog # PIPA569670
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of human VMDL3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BEST3 Bestrophin-3 VMD2L3 Bestrophin 3; Bestrophin-3; vitelliform macular dystrophy 2-like 3; Vitelliform macular dystrophy 2-like protein 3
Gene Aliases: BEST3; mBest4; VMD2L3
UniProt ID: (Human) Q8N1M1, (Mouse) Q6H1V1
Entrez Gene ID: (Dog) 481158, (Rabbit) 100350241, (Horse) 100052270, (Human) 144453, (Guinea pig) 100721635, (Mouse) 382427, (Rat) 314847
Molecular Function:
anion channel
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.