Invitrogen
Manufacturer Code:PA576810
Catalog # PIPA576810
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human Bestrophin 1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARB BEST 1 BEST Best disease Best macular dystrophy BEST1 BEST1&lowbarHUMAN Best1V1Delta2 Bestrophin 1 Bestrophin-1 Bestrophin1 BMD mBest1 RP50 TU15B Vitelliform macular dystrophy 2 Vitelliform macular dystrophy Vitelliform macular dystrophy protein 2 VMD 2 VMD2; Best disease; best macular dystrophy; Best1V1Delta2; Bestrophin-1; TU15B; vitelliform macular dystrophy 2 homolog; Vitelliform macular dystrophy protein 2; Vitelliform macular dystrophy protein 2 homolog
Gene Aliases: ARB; BEST; BEST1; BMD; Bmd1; mBest1; RP50; TU15B; VMD2
UniProt ID: (Human) Q8IUR9, (Mouse) O88870
Entrez Gene ID: (Human) 7439, (Mouse) 24115
Molecular Function: anion channel ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.