Novus Biologicals
Manufacturer Code:NBP292979100UL
Catalog # NB027779
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 376-585 of human BEST1 (NP_004174.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARB BEST Best disease bestrophin 1 bestrophin-1 BMD TU15B vitelliform macular dystrophy 2 Vitelliform macular dystrophy protein 2 VMD2RP50; Best disease; Best1V1Delta2; Bestrophin-1; TU15B; Vitelliform macular dystrophy protein 2
Gene Aliases: ARB; BEST; BEST1; BMD; RP50; TU15B; VMD2
UniProt ID: (Human) Q8IUR9
Entrez Gene ID: (Human) 7439
Molecular Function:
anion channel
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.