Invitrogen
Manufacturer Code:PA529334
Catalog # PIPA529334
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 907 and 1231 of Human Bcr |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ALL BCR1 CML D22S11 D22S662 PHL breakpoint cluster region protein renal carcinoma antigen NY-REN-26; BCR/FGFR1 chimera protein; breakpoint cluster region; Breakpoint cluster region protein; FGFR1/BCR chimera protein; Renal carcinoma antigen NY-REN-26
Gene Aliases: ALL; BCR; BCR1; CML; D22S11; D22S662; PHL
UniProt ID: (Human) P11274
Entrez Gene ID: (Human) 613
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.