Abnova
Manufacturer Code:MAB3894
Catalog # 89-111-649
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Chinese hamster |
Class | Monoclonal |
Type | Antibody |
Clone | B325H |
Immunogen | Recombinant 6His fusion protein corresponding to mouse Bcl3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Protein Aliases: B-cell CLL/lymphoma 3; B-cell leukemia/lymphoma 3; B-cell lymphoma 3 protein homolog; BCL-3
Gene Aliases: AI528691; Bcl-3; Bcl3
UniProt ID: (Mouse) Q9Z2F6
Entrez Gene ID: (Mouse) 12051
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.