Abnova
Manufacturer Code:MAB2077
Catalog # 89-110-811
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7D9/SPM519 |
Immunogen | A synthetic peptide corresponding to N-terminus of mouse Bcl2l1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Mouse monoclonal antibody raised against synthetic peptide of Bcl2l1.
For Research Use Only
Protein Aliases: anti-apoptosis regulatory protein; Apoptosis regulator Bcl-X; B-cell leukemia/lymphoma x; Bcl(X)L; Bcl-2-like protein 1; bcl-x; Bcl-XL; Bcl-Xs; Bcl2-L-1; Bcl2l; BclX; MGC113803; MGC99998
Gene Aliases: Bcl(X)L; bcl-x; Bcl-XL; bcl2-L-1; Bcl2l; Bcl2l1; BclX
UniProt ID: (Mouse) Q64373
Entrez Gene ID: (Mouse) 12048
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.