Novus Biologicals
Manufacturer Code:NBP2252400.2MG
Catalog # NBP22524002
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | BX006 |
Immunogen | A synthetic peptide aa 3-14 (Cys-QSNRELVVDFLS) of human Bcl-X protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. |
For Research Use Only
Protein Aliases: Apoptosis regulator Bcl-X; Apoptosis regulator Bcl-X bcl2-L-1 BCL2-like 1 bcl-2-like protein 1 BCLXBCL2LBcl-X bcl-xL BCLXL BCL-XL/S bcl-xS BCLXS DKFZp781P2092; Bcl-2-like protein 1; Bcl2-L-1; BCL2-like 1; protein phosphatase 1, regulatory subunit 52
Gene Aliases: Bcl-X; BCL-XL/S; BCL2L; BCL2L1; BCLX; PPP1R52
UniProt ID: (Human) Q07817
Entrez Gene ID: (Human) 598
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.