Invitrogen
Manufacturer Code:MA529655
Catalog # PIMA529655
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 36 |
Immunogen | Recombit Mouse BCL2L1/Bcl-XL Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: anti-apoptosis regulatory protein; Apoptosis regulator Bcl-X; B-cell leukemia/lymphoma x; Bcl-2-like protein 1; BCL-XL/S BCL2L BCLX Bcl-X DKFZp781P2092 bcl-xL bcl-xS; Bcl2-L-1
Gene Aliases: Bcl(X)L; bcl-x; Bcl-XL; bcl2-L-1; Bcl2l; Bcl2l1; BclX
UniProt ID: (Mouse) Q64373
Entrez Gene ID: (Mouse) 12048
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.