Invitrogen
Manufacturer Code:MA529049
Catalog # PIMA529049
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 16 |
Immunogen | Recombit Human BCL2L1/Bcl-XL Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Apoptosis regulator Bcl-X; Bcl-2-like protein 1; BCL-XL/S BCL2L BCLX Bcl-X DKFZp781P2092 bcl-xL bcl-xS; Bcl2-L-1; BCL2-like 1; protein phosphatase 1, regulatory subunit 52
Gene Aliases: Bcl-X; BCL-XL/S; BCL2L; BCL2L1; BCLX; PPP1R52
UniProt ID: (Human) Q07817
Entrez Gene ID: (Human) 598
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.