OriGene
Manufacturer Code:CF500097
Catalog # 50-165-9354
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4A9 |
Immunogen | Recombit fragment expressed in E.coli corresponding to amino acids 1-212 of human Bcl-XL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Apoptosis regulator Bcl-X; Bcl-2-like protein 1; BCL-XL/S BCL2L BCLX Bcl-X PPP1R52; Bcl2-L-1; BCL2-like 1; protein phosphatase 1, regulatory subunit 52
Gene Aliases: Bcl-X; BCL-XL/S; BCL2L; BCL2L1; BCLX; PPP1R52
UniProt ID: (Human) Q07817
Entrez Gene ID: (Human) 598
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.