Proteintech
Manufacturer Code:269671AP150UL
Catalog # 50-173-5517
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Bcl-XL Fusion Protein Ag25604 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: anti-apoptosis regulatory protein; Apoptosis regulator Bcl X BCL2L1 Bcl-xL; Apoptosis regulator Bcl-X; B-cell leukemia/lymphoma x; Bcl-2-like protein 1; Bcl2-L-1; BCL2-like 1; protein phosphatase 1, regulatory subunit 52
Gene Aliases: Bcl(X)L; Bcl-X; Bcl-XL; BCL-XL/S; bcl2-L-1; BCL2L; BCL2L1; BCLX; PPP1R52
UniProt ID: (Human) Q07817, (Mouse) Q64373
Entrez Gene ID: (Human) 598, (Mouse) 12048
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.