Invitrogen
Manufacturer Code:PA146479
Catalog # PIPA146479
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues 1-100 of an N-terminal portion of rat BclX. &lsqbSwiss-ProtNo. Q07817] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: anti-apoptotic Bcl-2 gene family member; Apoptosis regulator Bcl X Bcl 2 like 1 Bcl 2 like 1 protein Bcl xL BCL XL/S ; Apoptosis regulator Bcl-X; Bcl-2-like protein 1; Bcl2-L-1; BCL2-like 1; BCL2L1; protein phosphatase 1, regulatory subunit 52
Gene Aliases: Bcl-X; BCL-XL; BCL-XL/S; BCL2L; BCL2L1; BCLX; PPP1R52
UniProt ID: (Human) Q07817
Entrez Gene ID: (Human) 598, (Dog) 403618, (Sheep) 443061, (Cat) 493701
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.