Invitrogen
Manufacturer Code:MA531647
Catalog # PIMA531647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 8A2F9 |
Immunogen | Purified recombit fragment of human BCL2L10 (AA: 31-186) expressed in E. coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Anti-apoptotic protein NrH; Apoptosis regulator Bcl-B; Bcl-2-like protein 10; Bcl2-L-10; BCL2-like 10 (apoptosis facilitator); Boo Diva BCL-B bcl2-L-10; death inducer binding to vBcl-2 and Apaf-1
Gene Aliases: BCL-B; bcl2-L-10; BCL2L10; BCLB; Boo; Diva
UniProt ID: (Human) Q9HD36
Entrez Gene ID: (Human) 10017
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.