Invitrogen
Manufacturer Code:MA517670
Catalog # PIMA517670
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Armenian hamster |
Class | Monoclonal |
Type | Antibody |
Clone | Ham 150-3.5 |
Immunogen | recombit Bcl-3-6-histidine tag |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Armenian Hamster Monoclonal Antibody
For Research Use Only
Protein Aliases: B-cell CLL/lymphoma 3; B-cell leukemia/lymphoma 3; B-cell lymphoma 3 protein homolog; B-cell lymphoma 3-encoded protein Bcl-3 protein; BCL-3
Gene Aliases: AI528691; Bcl-3; Bcl3
UniProt ID: (Mouse) Q9Z2F6
Entrez Gene ID: (Mouse) 12051
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.