Novus Biologicals
Manufacturer Code:NBP271398G
Catalog # NBP271398G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4A8 |
Immunogen | Full length recombinant protein of human Bcl-10 (NP_003912) produced in HEK293T cells. |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: B-cell CLL/lymphoma 10; B-cell CLL/lymphoma 10CIPERCLAPB-cell lymphoma/leukemia 10 Bcl-10 CARD-containing apoptotic signaling protein CARD-containing molecule enhancing NF-kappa-B CARD-containing proapoptotic protein CARD-like apoptotic protein CARMEN caspase-recruiting domain-containing protein cCARMEN c-E10bcl-10 CED-3/ICH-1 prodomain homologous E10-like regulator Cellular homolog of vCARMEN cellular-E10 hCLAP Mammalian CARD-containing adapter molecule E10 mE10CARD containing molecule enhancing NF-kB; B-cell lymphoma/leukemia 10; Bcl-10; c-E10; CARD containing molecule enhancing NF-kB; CARD-containing apoptotic signaling protein; CARD-containing molecule enhancing NF-kappa-B; CARD-containing proapoptotic protein; CARD-like apoptotic protein; caspase-recruiting domain-containing protein; cCARMEN; CED-3/ICH-1 prodomain homologous E10-like regulator; Cellular homolog of vCARMEN; Cellular-E10; CIPER; hCLAP; Mammalian CARD-containing adapter molecule E10; mE10
Gene Aliases: BCL10; c-E10; CARMEN; CIPER; CLAP; IMD37; mE10
UniProt ID: (Human) O95999
Entrez Gene ID: (Human) 8915
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.