Invitrogen
Manufacturer Code:PA524191
Catalog # PIPA524191
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 24-51 amino acids from the N-terminal region of human BCL10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B-cell CLL/lymphoma 10; B-cell leukemia/lymphoma 10; B-cell lymphoma/leukemia 10; Bcl-10; c-E10; CARD containing molecule enhancing NF-kB; CARD-containing apoptotic signaling protein; CARD-containing molecule enhancing NF-kappa-B; CARD-containing proapoptotic protein; CARD-like apoptotic protein; CARMEN CIPER CLAP c-E10 mE10 B-cell lymphoma/leukemia 10 CARD containing molecule enhancing NF-kB CARD-containing apoptotic signaling protein CARD-containing molecule enhancing NF-kappa-B CARD-containing proapoptotic protein CED-3/ICH-1 prodomain homologous E10-like regulator cCARMEN; caspase-recruiting domain-containing protein; cCARMEN; CED-3/ICH-1 prodomain homologous E10-like regulator; Cellular homolog of vCARMEN; Cellular-E10; CIPER; hCLAP; Mammalian CARD-containing adapter molecule E10; mCIPER; mCLAP; mE10
Gene Aliases: AI132454; BCL-10; BCL10; c-E10; C81403; CARMEN; cE10; CIPER; CLAP; IMD37; ME10
UniProt ID: (Human) O95999, (Mouse) Q9Z0H7
Entrez Gene ID: (Human) 8915, (Mouse) 12042
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.