Novus Biologicals
Manufacturer Code:NBP224912FR
Catalog # NBP224912FR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 20F565 |
Immunogen | A synthetic peptide made to the N-terminal portion of the human Bax inhibitor 1 protein (between amino acids 1-40) UniProt P55061. |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Bax inhibitor 1; BAXI1 BI1 BI-1Transmembrane BAX inhibitor motif-containing protein 6 TEGTbax inhibitor 1 testis enhanced gene transcript Testis-enhanced gene transcript protein transmembrane BAX inhibitor motif containing 6; BI-1; testis enhanced gene transcript; Testis-enhanced gene transcript protein; Transmembrane BAX inhibitor motif-containing protein 6
Gene Aliases: BAXI1; BI-1; BI1; TEGT; TMBIM6
UniProt ID: (Human) P55061
Entrez Gene ID: (Human) 7009
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.