Novus Biologicals
Manufacturer Code:NB10056095
Catalog # NB10056095
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 43-61 (PELALDPVPQDASTKKLSE) of human Bax alpha was used as immunogen GenBank no. NP_620116.1. The immunogen sequence is 100% conserved in human Bax isoforms alpha (192 amino acids) beta (218 amino acids) epsilon (164 amino acids) sigma (179 amino acids) and psi (173 amino acids). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Apoptosis regulator BAX; apoptosis regulator BAX BCL2-associated X protein Bcl2-L-4 BCL2L4bcl2-L-4 Bcl-2-like protein 4; Baxdelta2G9; Baxdelta2G9omega; Baxdelta2omega; Bcl-2-like protein 4; BCL2 associated X protein; BCL2-associated X protein omega; Bcl2-L-4
Gene Aliases: BAX; BCL2L4
UniProt ID: (Human) Q07812
Entrez Gene ID: (Human) 581
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.