Invitrogen
Manufacturer Code:PA5114895
Catalog # PIPA5114895
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Bassoon corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bassoon (presynaptic cytomatrix protein); bassoon bassoon (presynaptic cytomatrix protein) bassoon presynaptic cytomatrix protein BSN Kiaa0434 neuronal double zinc finger protein presynaptic cytomatrix protein protein bassoon Zinc finger protein 231 ZNF231; neuronal double zinc finger protein; presynaptic cytomatrix protein; Protein bassoon; Zinc finger protein 231
Gene Aliases: BSN; KIAA0434; ZNF231
UniProt ID: (Human) Q9UPA5, (Mouse) O88737
Entrez Gene ID: (Human) 8927, (Rat) 29138, (Mouse) 12217
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.