Novus Biologicals
Manufacturer Code:NBP298703R
Catalog # NB054778
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Band 3. |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AE 1; AE 1 AE1MGC126619 Anion exchange protein 1 Anion exchanger 1 band 3 anion transport protein BND3 CD233 CD233 antigen DI EMPB3 EPB3MGC126623 erythrocyte membrane protein band 3 erythroid anion exchange protein FR Froese blood group MGC116750 MGC116753 RTA1A Solute carrier family 4 member 1 solute carrier family 4 anion exchanger member 1 (erythrocyte membraneprotein band 3 Diego blood group) solute carrier family 4 anion exchanger number 1 SW Swann blood group Waldner blood group WD WD1 WR Wright blood group; Anion exchange protein 1; anion exchanger 1; anion exchanger-1; Band 3 anion transport protein; CD233; Diego blood group; erythrocyte membrane protein band 3; erythroid anion exchange protein; Froese blood group; solute carrier family 4 (anion exchanger), member 1 (Diego blood group); Solute carrier family 4 member 1; solute carrier family 4, anion exchanger, member 1 (erythrocyte membrane protein band 3, Diego blood group); solute carrier family 4, anion exchanger, number 1; Swann blood group; Waldner blood group; Wright blood group
Gene Aliases: AE1; BND3; CD233; CHC; DI; EMPB3; EPB3; FR; RTA1A; SAO; SLC4A1; SPH4; SW; WD; WD1; WR
UniProt ID: (Human) P02730
Entrez Gene ID: (Human) 6521
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.