R&D Systems
Manufacturer Code:MAB852SP
Catalog # MAB852SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | GP3.10G3E2 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Bag-1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BAG family molecular chaperone regulator 1; BAG family molecular chaperone regulator 1 Bag1 BAG-1 Bcl-2 associating athanogene-1 protein BCL2-associated athanogene Bcl-2-associated athanogene 1 Bcl-2-binding protein glucocortoid receptor-associated protein RAP4646-KD HAP RAP46; BAG-1; Bcl-2 associating athanogene-1 protein; Bcl-2-associated athanogene 1; Bcl-2-binding protein; BCL2-associated athanogene; glucocortoid receptor-associated protein RAP46; receptor-associated protein, 46-KD
Gene Aliases: BAG-1; BAG1; HAP; RAP46
UniProt ID: (Human) Q99933
Entrez Gene ID: (Human) 573
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.