Novus Biologicals
Manufacturer Code:NB10056083
Catalog # NB10056083
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Full-length recombinant mouse BAG-1 protein was used as immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BAG family molecular chaperone regulator 1; BAG family molecular chaperone regulator 1 BAG-1 Bcl-2 associating athanogene-1 protein BCL2-associated athanogene Bcl-2-associated athanogene 1 Bcl-2-binding protein glucocortoid receptor-associated protein RAP4646-KD HAP RAP46; BAG-1; Bcl-2 associating athanogene-1 protein; Bcl-2-associated athanogene 1; Bcl-2-binding protein; BCL2-associated athanogene; glucocortoid receptor-associated protein RAP46; receptor-associated protein, 46-KD
Gene Aliases: BAG-1; BAG1; HAP; RAP46
UniProt ID: (Human) Q99933
Entrez Gene ID: (Human) 573
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.