Novus Biologicals
Manufacturer Code:NB10056080
Catalog # NB10056080
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A full length recombinant protein corresponding to human Bad was used as immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BAD; BBC2 BBC6 bcl2 antagonist of cell death BCL2-antagonist of cell death protein BCL2-associated agonist of cell death Bcl-2-binding component 6 BCL2-binding component 6 BCL2-binding protein Bcl2-L-8 BCL2L8bcl2-L-8 Bcl-2-like protein 8 BCL-X/BCL-2 binding protein Bcl-XL/Bcl-2-associated death promoter; Bcl-2-binding component 6; Bcl-2-like protein 8; BCL-X/BCL-2 binding protein; Bcl-xL/Bcl-2-associated death promoter; Bcl2 antagonist of cell death; BCL2-antagonist of cell death protein; BCL2-associated agonist of cell death; BCL2-binding component 6; BCL2-binding protein; Bcl2-L-8
Gene Aliases: BAD; BBC2; BBC6; BCL2L8
UniProt ID: (Human) Q92934
Entrez Gene ID: (Human) 572
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.