Invitrogen
Manufacturer Code:PA555145
Catalog # PIPA555145
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human BZRAP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: benzodiazapine receptor (peripheral) associated protein 1; benzodiazapine receptor peripheral-associated protein 1; benzodiazepine receptor (peripheral) associated protein 1; PBR-IP; Peripheral benzodiazepine receptor-interacting protein; Peripheral-type benzodiazepine receptor-associated protein 1; Peripheral-type benzodiazepine receptor-associated protein 1 PRAX-1 Peripheral benzodiazepine receptor-interacting protein PBR-IP RIMS-binding protein 1 RIM-BP1 BZRAP1 KIAA0612 RBP1 RIMBP1; PRAX-1; RIM-BP1; RIMS-binding protein 1; TSPO-associated protein 1
Gene Aliases: BZRAP1; KIAA0612; PBR-IP; PRAX-1; PRAX1; RBP1; RIM-BP1; RIMBP1; TSPOAP1
UniProt ID: (Human) O95153
Entrez Gene ID: (Human) 9256
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.