Novus Biologicals
Manufacturer Code:NBP193626
Catalog # NBP193626
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:DPGAMEPWALPTWHSWTPGRGGEPSSAAPSIADTPPAALQLQELRSEESSKPKGDGSSRPVGGTDPEGAEACLPSLGQQASSSGPACQRPEDEEVEAFLKAKLNMSFGD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: benzodiazapine receptor (peripheral) associated protein 1; benzodiazapine receptor (peripheral) associated protein 1 DKFZp686F02123 KIAA0612 PBR-IP peripheral benzodiazepine receptor-associated protein 1 peripheral benzodiazepine receptor-interacting protein peripheral-type benzodiazepine receptor-associated protein 1 PRAX1 PRAX-1 RBP1 RIMBP1 RIM-BP1 RIMS-binding protein 1; benzodiazapine receptor peripheral-associated protein 1; benzodiazepine receptor (peripheral) associated protein 1; PBR-IP; Peripheral benzodiazepine receptor-interacting protein; Peripheral-type benzodiazepine receptor-associated protein 1; PRAX-1; RIM-BP1; RIMS-binding protein 1; TSPO-associated protein 1
Gene Aliases: BZRAP1; KIAA0612; PBR-IP; PRAX-1; PRAX1; RBP1; RIM-BP1; RIMBP1; TSPOAP1
UniProt ID: (Human) O95153
Entrez Gene ID: (Human) 9256
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.