Invitrogen
Manufacturer Code:PA541912
Catalog # PIPA541912
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human BXDC5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310066N05Rik; brix domain containing 5; Brix domain-containing protein 5; Ribosome biogenesis protein RPF1; Ribosome production factor 1; ribosome production factor 1-like protein; RNA processing factor 1; RNA processing factor 1 (RPF1)
Gene Aliases: 2210420E24Rik; 2310066N05Rik; BXDC5; RPF1
UniProt ID: (Human) Q9H9Y2, (Mouse) Q7TND5
Entrez Gene ID: (Human) 80135, (Dog) 479972, (Bovine) 513081, (Rabbit) 100341955, (Horse) 100064844, (Mouse) 70285, (Guinea pig) 100736070, (Rat) 499725
Molecular Function: RNA binding protein nucleic acid binding ribonucleoprotein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.