Novus Biologicals
Manufacturer Code:NBP189915
Catalog # NBP189915
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:SGKKSLKRKAAAEELQEAAGAGDGATENGVQPPKAAAFPPGFSISEIKNKQRRHLMFTRWKQQQRKEKLAAKKKLKKEREALGDKAPPKPVPKTID |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310066N05Rik; 2310066N05Rik BBR140 brix domain containing 5 Brix domain-containing protein 5 BXDC5 DKFZp761G0415 DKFZp761M0215 FLJ12475 Ribosome biogenesis protein RPF1 ribosome production factor 1 ribosome production factor 1 homolog (S. cerevisiae) RNA processing factor 1 (RPF1) RP11-118B23.1; brix domain containing 5; Brix domain-containing protein 5; Ribosome biogenesis protein RPF1; Ribosome production factor 1; RNA processing factor 1 (RPF1)
Gene Aliases: BXDC5; RPF1
UniProt ID: (Human) Q9H9Y2
Entrez Gene ID: (Human) 80135
Molecular Function: RNA binding protein nucleic acid binding ribonucleoprotein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.