Proteintech
Manufacturer Code:172951AP150UL
Catalog # 50-173-1495
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | BXDC2 Fusion Protein Ag11174 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BRIX brix domain containing 2 BRIX1 BXDC2; brix domain containing 2; Brix domain-containing protein 2; BRX1, biogenesis of ribosomes, homolog; ribosome biogenesis protein Brix; Ribosome biogenesis protein BRX1 homolog
Gene Aliases: 1110064N10Rik; BRIX; BRIX1; BXDC2; C76935; RGD1308508
UniProt ID: (Human) Q8TDN6, (Mouse) Q9DCA5, (Rat) Q4QQT6
Entrez Gene ID: (Human) 55299, (Mouse) 67832, (Rat) 294799
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.